Middlebrook 7H11 Selective HardyFlask™, Hardy Diagnostics
Catalog # 75814-682
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:Middlebrook 7H11 Selective HardyFlask™
- Size:Pack of 20
- Cat. no.:75814-682
- Volume:12 ml
- Supplier No.:X28
Specifications
About this item
Middlebrook 7H11 Selective is used for selective isolation of Mycobacterium spp.
- Mitchison formula
- CAPT antibiotics
- Selective isolation
Mitchison formula with CAPT antibiotics, for selective isolation of Mycobacterium spp.