Order Entry
Canada
ContactUsLinkComponent
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
Catalog # 75813-960
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Lauryl Tryptose Broth with MUG (Durham Tube), Hardy Diagnostics
Catalog # 75813-960
Supplier:  Hardy Diagnostics
Supplier Number:  K238
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Lauryl Tryptose w/MUG
  • Size:
    Pack of 20
  • Minimum Order Quantity:
    60
  • Cat. no.:
    75813-960
  • Format:
    20×125 mm tube
  • Supplier No.:
    K238

Specifications

About this item

Recommended as a test of Escherichia coli and coliforms by fermentation with gas production and fluorogenic means

  • Fluorogenic test means
  • Test of Escherichia coli and coliforms
  • Gas production and MUG reaction detection

For the detection of E. coli by gas production and MUG reaction.