You searched for: Standard Laboratory Blenders
VWR’s laboratory blenders are powerful and durable for grinding, blending, chopping, and processing. Designed specifically with the daily requirements of a lab in mind, standard blenders are available in either stainless steel or glass, with solid bases of support in all models. Safe and easy to clean, these laboratory blenders are used with both wet and dry ingredients, and are available in various sizes and speed settings.
Hazardous Location Blender Motor, Waring
Supplier: Waring Products
Waring® blenders to provide optimum safety and reliable performance when working with highly volatile substances.
Expand 1 Items
Single-Speed Blenders, 1 L, Waring
Supplier: Waring Products
For rapid pulping or mixing
Expand 2 Items
Dry Blending Container, Waring
Supplier: Waring Products
Unique blade configuration designed for pulverizing and homogenizing dry substances
Expand 1 Items
Two-Speed Laboratory Blenders, 1 L, Waring
Supplier: Waring Products
Versatile two-speed blenders (18,000 and 22,000rpm) for general laboratory work.
Expand 2 Items
Two-Speed Lab Blenders, Waring
Supplier: Waring Products
This versatile laboratory blender is applicable for most lab work, providing no-load speeds of 18,000rpm and 22,000rpm
Expand 1 Items
Stainless Steel Container for Waring® Blenders, 360 ml, Eberbach
Supplier: Eberbach
Special purpose container for rapidly disintegrating and mixing small volumes of microorganisms and chemicals
Expand 2 Items
Blending Assembly, Waring
Supplier: Waring Products
For one and two liter Waring® blenders.
Expand 1 Items
Stainless Steel Mini-Containers for 1 L Blenders, Waring
Supplier: Waring Products
For processing small quantities
Expand 3 Items
Stainless Steel Containers for Blenders, Waring
Supplier: Waring Products
1L. With one-piece stainless steel lid. Standard blending assembly with fluoropolymer resin gaskets. Fits all 1L Waring* laboratory blenders.
Expand 2 Items
Glass Container for Waring® Blenders, 500 ml, Eberbach
Supplier: Eberbach
For use where smaller quantities of material are desired
Expand 2 Items
20 ml Stainless Steel Micro Container for Waring® Blenders, Eberbach
Supplier: Eberbach
Working capacity 3–20mL
Expand 1 Items
Variable Speed Laboratory Blender, Waring
Supplier: Waring Products
Variable speed blender allows adjustment of speeds for mixing, stirring, blending, or homogenizing
Expand 2 Items
Three-Speed Commercial Blender, 4 L, Waring
Supplier: Waring Products
Designed for high speed blending of large quantities of highly liquid material.
Expand 1 Items
Pulverizer, Waring
Supplier: Waring Products
Small stainless steel container with special blade configurations reduces small bones, seeds, grains, nuts, soybeans, tablets, soils and similar materials in seconds
Expand 1 Items
Stainless Steel Container for Waring® Blenders, Eberbach
Supplier: Eberbach
Special purpose container assembly with stainless steel screw cover for sealing in contents and 15.9mm (5/8") threaded sampling plug for removal or addition of material