VWR® Bohler Double-Action Bone Cutting Forceps
Supplier: VWR International
Certificates
About this item
The VWR® Bohler Double-Action Bone Cutting Forceps are made from ASTM (899-94) Series 400 German stainless steel
- Passivated
- Surgical grade
- Rockwell Hardness (HRC): 50–55
- Compatible with Gamma/ETO/Autoclavable sterilization methods
Available in a straight or angled format, these instruments are dsigned for cutting cartilage and bone. A strong, removable bar-spring makes these shears self-opening.
Frequently Bought Together