ContactUsLinkComponent
VWR® Dissecting Bone Cutting Forceps
VWR® Dissecting Bone Cutting Forceps
Catalog # 10806-298
Supplier:  VWR International
undefined
VWR® Dissecting Bone Cutting Forceps
Catalog # 10806-298
Supplier:  VWR International
Specifications
  • Description:
    VWR® Dissecting Bone Cutting Forceps
  • Length:
    127 mm (5")
  • Material:
    Stainless steel, surgical grade
  • Cat. no.:
    10806-298
Specifications
About this item
Surgical Grade Stainless Steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods