ContactUsLinkComponent
Acanthamoeba Broth (PYG), for Free-Living Amebae Such as Acanthamoeba, Hardy Diagnostics
  10324-456
Product available on GSA Advantage®
 :  
undefined
Acanthamoeba Broth (PYG), for Free-Living Amebae Such as Acanthamoeba, Hardy Diagnostics
  10324-456
 :  K225
Product available on GSA Advantage®
 :  
  • Description:
    Acanthamoeba Broth (PYG), for Free-Living Amebae Such as Acanthamoeba
  • Size:
    5 mL
  • Minimum Order Quantity:
    10
  • Cat. no.:
    10324-456
  • Format:
    15×103 mm PC tube
  • Supplier No.:
    K225
Acanthamoeba Broth (PYG) is used for the maintenance of stock cultures.
  • Easy read-out
  • Simple procedure
  • Microscopically visualize distinct feeding tracks
Acanthamoeba Broth (PYG Medium) and Naegleria Broth are broths for the maintenance of stock cultures.
Non-Nutrient agar and Page's Saline are recommended for the detection of free-living pathogenic Acanthamoeba and Naegleria spp. cysts, trophozoites, and flagellates in tissue, soil, or water samples.
Simple procedure, prepared, ready-to-use media. Easy Read-out, microscopically visualize (100x) distinct feeding tracks left behind by amebae.
 : Minimum order of 10.