Sabdex (Sabouraud Dextrose) Agar, Hardy Diagnostics
Catalog # 10324-418
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:Sabdex (Sabouraud Dextrose) Agar
- Size:12 mL
- Cat. no.:10324-418
- Format:50 ml plastic HardyFlask™
- Supplier No.:X40
Specifications
About this item
Sabdex (Sabouraud Dextrose) Agar is used for fungi cultivation.
- Cultivation of fungi
For the cultivation of fungi.