ContactUsLinkComponent
Blood, Sheep, Defibrinated, Hardy Diagnostics
Blood, Sheep, Defibrinated, Hardy Diagnostics
Catalog # 10145-962
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
undefined
Blood, Sheep, Defibrinated, Hardy Diagnostics
Catalog # 10145-962
Supplier:  Hardy Diagnostics
Supplier Number:  SB50
Product available on GSA Advantage®
Specifications
  • Description:
    Blood, Sheep, Defibrinated
  • Size:
    50 ml
  • Cat. no.:
    10145-962
  • Supplier No.:
    SB50
Specifications
About this item
The sheep are humanely bled and rotated on a veterinary supervised program.
  • Derived form a donor herd
  • Antibiotic-free
  • Less prone to spontaneous hemolysis
This product has a short shelf life and ships from our California warehouse only.
Packaged in a glass bottle with a screw-cap. Our sheep blood is derived form a donor herd that is kept solely for Hardy Diagnostics. The sheep are humanely bled and rotated on a veterinary supervised program. In this way, the blood is kept antibiotic-free and is less prone to spontaneous hemolysis.