Round Cover Glass, Azer Scientific
Catalog #: 89187-782
Supplier: Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
Specifications
- Ø:12 mm
- Numbering:#1
- Description:Cover glasses, round
- Supplier No.:200121
Specifications
About this item
The round cover glass is pure white borosilicate glass of the first hydrolytic class.
- Excellent chemical resistance
- Optimal flatness
- No disrupting fluorescence
Cover glass does not emit any disturbing fluorescence under the ultraviolet light of the fluorescence microscope.
Customers Who Bought This Also Bought