Order Entry
Canada
ContactUsLinkComponent
Mikulicz Forceps, OR Grade, Sklar
Mikulicz Forceps, OR Grade, Sklar
Catalog # 10199-136
Supplier:  Sklar
undefined
Mikulicz Forceps, OR Grade, Sklar
Catalog # 10199-136
Supplier:  Sklar
Supplier Number:  90-2584

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Length:
    203 mm (8")
  • Material:
    Stainless steel
  • Curvature:
    Strong Curve
  • Cat. no.:
    10199-136
  • Tip Style:
    Serrated Jaws
  • Supplier No.:
    90-2584

Specifications

About this item

Mikulicz Forceps are constructed of stainless steel

Industry standard.