If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Length:197 mm (7¾")
- Material:Stainless Steel with Reduced Reflectivity Polymer Coating
- Curvature:Straight
- Cat. no.:10199-114
- Tip Style:1 x 2 Rows of Nontraumatic Fine Teeth
- Supplier No.:90-2310
Sklar Black™ DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used for grasping fine tissue during laser surgeries
They are commonly used in vascular and general surgery. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements.