Orders LinkContactUsLinkComponent
Knapp Iris Scissors, OR Grade, Sklar
Knapp Iris Scissors, OR Grade, Sklar
Supplier:   Sklar
Knapp Iris Scissors, OR Grade, Sklar
Supplier:  Sklar
Certificates
Knapp Iris Scissors, OR Grade, Sklar
Knapp Iris Scissors, OR Grade, Sklar
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Knapp Iris Scissors are a delicate ophthalmic instrument used for cutting and dissecting the iris.
Censitrac Ready products are industry standard.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Product Family Options
Knapp Iris Scissors, OR Grade
Product Information
  • Length
    Material
    Blade type
    Curvature
    Handle
    Sterility
    Availability
    Price
  • 10197-886
    Knapp Iris Scissors, Premium OR-Grade
    Knapp Iris Scissors, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $286.64
    $286.64
    New Customer Promotional Price
    Each
       $286.64
    Supplier Number: 64-3230
  • 10197-888
    Knapp Iris Scissors, Premium OR-Grade
    Knapp Iris Scissors, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Blunt/Blunt, Smooth
    Curved
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $284.11
    $284.11
    New Customer Promotional Price
    Each
       $284.11
    Supplier Number: 64-3235
  • 10197-902
    Knapp Iris Scissors, Premium OR-Grade
    Knapp Iris Scissors, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Sharp/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $261.47
    $261.47
    New Customer Promotional Price
    Each
       $261.47
    Supplier Number: 64-3430
  • 10197-904
    Knapp Iris Scissors, Premium OR-Grade
    Knapp Iris Scissors, Premium OR-Grade
    10.2 cm (4")
    Stainless steel
    Blunt/Blunt, Smooth
    Straight
    Finger Rings
    Non sterile
    Log in for availability
    Log in for availability
    Each
    Each - $261.47
    $261.47
    New Customer Promotional Price
    Each
       $261.47
    Supplier Number: 64-3435
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)