This activity encourages students to use deductive reasoning when analyzing unknowns.
- Students Conduct Experiments and Identify Chemical Reactions
- Activities Include Mixing Seven Solutions into 21 Possible Combinations to Observe Reactions
- Purpose: to Help Students Use Deductive Reasoning & Standard Lab Procedures
- Chemical Reactions: Color Changes, Precipitates, Gas Generation & More
- Extension Assignment: Probable Chemical Equation of Each Reaction
Students first mix seven known solutions into any 21 possible combinations, using standard lab procedures. They then observe and make notations as to how the chemicals react.
After recording their observations, students must decide whether a chemical reaction has taken place and provide evidence to support their deductions. As an extension, students can write the probable chemical equation of each reaction.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.