Orders LinkContactUsLinkComponent
Introduction to Forensics Analysis
Introduction to Forensics Analysis
Supplier:   Ward's Science
Introduction to Forensics Analysis
Supplier:  Ward's Science
Certificates
Introduction to Forensics Analysis
Introduction to Forensics Analysis
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Introduce Students to an Array of Forensics Fields
  • Five lab activities in one bundle
This kit pack includes the necessary materials to introduce students to the forensic fields of hair analysis, fingerprint analysis, soil and mineral analysis, drug and poison analysis and document analysis.
Product Family Options
Product Information
  • Includes
    No. of lab groups
    Availability
    Price
  • 470220-222
    Introduction to Forensics Analysis
    Introduction to Forensics Analysis
    Teacher ‘s Guide and Student Copymasters
    10
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $1,045.00
    $1,045.00
    Kit of 1
       $1,045.00
    Supplier Number: IS9020
  • 470237-792
    Introduction to Forensics Analysis, Refill Kit
    Introduction to Forensics Analysis, Refill Kit
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $1,027.65
    $1,027.65
    Kit of 1
       $1,027.65
    Supplier Number: IS9020-REF
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)