ContactUsLinkComponent
Green Fuel Cell Lab Activity
Green Fuel Cell Lab Activity
Catalog # 470221-558
Supplier:  Ward's Science
undefined
Green Fuel Cell Lab Activity
Catalog # 470221-558
Supplier:  Ward's Science
Supplier Number:  IS6500
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Green Fuel Cell Lab Activity
  • No. of Lab Groups:
    15
  • Cat. no.:
    470221-558
  • Supplier No.:
    IS6500
Specifications
About this item
The Green Fuel Cell Lab Activity is a great introduction to green energy production.
  • Users Monitor the Voltage Generated by a Biological Fuel Cell
  • Ideal Introduction to Green Energy Production
  • Methylene Blue Works as an Electron Mediator
Create a biological fuel cell using the yeast Saccharomyces cerevisiae. A simple sugar is used as a food source for the yeast, and methylene blue is employed as an electron mediator. The cell is constructed without the mediator, and a baseline voltage is established as some electrochemical activity occurs.
Once the cell has stabilized, the mediator is added, and the effect on voltage is monitored and recorded using a voltage sensor. A voltage sensor is required.
Ordering information: This kit includes dialysis tubing, glucose, yeast, methylene blue, potassium ferricyanide, phosphate buffer, red connecting wires, black connecting wires, electrodes, and filter paper. A voltage sensor is required but not included.