If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Effectiveness of Sunscreen
- No. of Lab Groups:15
- Cat. no.:470221-562
- Supplier No.:IS6502
Specifications
About this item
This activity illustrates the importance sunscreen use.
- For Use with NeuLog Logger Sensors
- Students See the Importance of Using Sunscreen
- Kit Provides Microscope Slides and SPF 15 and 30
- Teacher's Guide and Student Copy Master Included
Electromagnetic radiation in the UV range of the spectrum cannot be perceived by the human eye. Using the NeuLog UVB sensor specifically designed to measure UVB radiation, users compare the amount of UVB radiation reaching the sensor.
The sensor is covered by both SPF 15 and SPF 30 sunscreens to determine if the sunscreens actually provide UVB protection, and if so, how much protection.