SP Wilmad-LabGlass Laboratory Grade Automatic Burettes with PTFE Plug, SP Industries
Supplier: SP Industries
Certificates
About this item
This inexpensive portable titration set features a 2mm straight stopcock with PTFE plug
The borosilicate glass burette is filled by pressure supplied by the rubber bulb. A small hole in the filling tube at the "O" line permits setting of the liquid level and excess titrant is returned to the bottle. Complete Set furnished with burette, bottle, rubber hose, rubber stopper, and rubber bulb.
Frequently Bought Together