Anti-CST8 Rabbit Polyclonal Antibody
100 μg
Catalog # 10792-382
Supplier: Genetex
Supplier #: GTX53934
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Rabbit polyclonal antibody to Cystatin 8
Type: Primary
Antigen: CST8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Size:100 μg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity purification
- Antibody type:Primary
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Store at –20 °C (regular) or –80 °C (long term). Avoid freeze / thaw cycles.
- Host:Rabbit
- Antigen name:Cystatin 8
- Cat. No.:10792-382
- Reactivity:Human|Mouse
- Gene ID:10047
- Antigen synonyms:CRES|CTES5
- Immunogen:Recombinant protein of human CST8
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:CST8
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH7.3.
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



