Anti-PCID2 Rabbit Polyclonal Antibody
Catalog # 10165-002
Supplier: Genetex
Specifications
- Antibody type:Primary
- Antigen name:Pci Domain Containing 2
- Antigen symbol:PCID2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 μg
- Gene ID:55795
- Antigen synonyms:F10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and less than 0.09% sodium azide.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:KLH conjugated synthetic peptide derived from human PCID2
- Purification:Purified by Protein A and peptide affinity chromatography.
- Cat. no.:10165-002
- Supplier No.:GTX52023
Specifications
About this item
Rabbit Polyclonal antibody to PCID2
Type: Primary
Antigen: PCID2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat