Order Entry
Canada
ContactUsLinkComponent
Ammonium Standard for Ion chromatography (IC), Certipur®, MilliporeSigma
Ammonium Standard for Ion chromatography (IC), Certipur®, MilliporeSigma
  CA1.19812.0500
 :  
Ammonium Standard for Ion chromatography (IC), Certipur®, MilliporeSigma
  CA1.19812.0500
 :  1.19812.0500
 :  

 

  • Concentration:
    1000 ppm
  • Pack type:
    Plastic Bottle
  • Description:
    Ammonium standard solution
  • Size:
    500 ml
  • Hazard class description:
    Corrosive Materials
  • Cat. no.:
    CA1.19812.0500
  • pH:
    5.4 (H₂O, 20°C)
  • Contents:
    NH₄Cl in H₂O
  • Supplier No.:
    1.19812.0500

 

 

Traceable to SRM from NIST.