Spill-X-S® Solvent Absorbent, PIG®
Catalog #: 10860-630
Supplier: 2 D 2 S.A. DE C.V. MX
Specifications
- Description:Spill-X-S® Solvent Absorbent
- Cat. no.:10860-630
- Absorbency:3 gal.
- Supplier No.:GEN324
Specifications
About this item
Absorbs larger spills and decreases the chance of ignition for safer cleanup and handling.
- Adsorbs spills of many common hydrocarbon solvents, reagents and fuels; solidified liquid is easy to clean up
- Granular base lets you use only what you need; reduces product waste and disposal costs
- Decreases chance of ignition by elevating flashpoint of solvent to above 140 ºF
- Speeds spill cleanup to limit worker exposure and minimize production downtime
- Non-toxic, free-flowing dry chemical is environmentally friendly
Ideal for use in chemical plants, manufacturing facilities, laboratories, vehicles transporting organic solvents or fuels and anywhere that is prone to solvent spills
Customers Who Bought This Also Bought