Order Entry
Canada
ContactUsLinkComponent
Tellurium Standard for AAS, Certipur®, MilliporeSigma
Tellurium Standard for AAS, Certipur®, MilliporeSigma
Catalog # CA1.19514.0100
Supplier:  MilliporeSigma
CAS Number:  
Tellurium Standard for AAS, Certipur®, MilliporeSigma
Catalog # CA1.19514.0100
Supplier:  MilliporeSigma
Supplier Number:  1.19514.0100
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard class description:
    Corrosive Materials
  • Cat. no.:
    CA1.19514.0100
  • pH:
    0.5 (H₂O, 20°C)
  • Element:
    Tellurium
  • Supplier No.:
    1.19514.0100

Specifications

About this item

Traceable to SRM from NIST.