Raw Polyurethane Foam (PUF) Plugs, Restek
Supplier: Restek
Certificates
About this item
Unwashed PUF plugs for both low-volume and high-volume sampling.
- Flame retardant free
- Easier to clean for trace analysis
- Use for collection of semivolatiles (pesticides, PCBs, PAHs)
- Suitable for ambient, indoor, and industrial hygiene applications
Both large high-volume (220 to 280 l/min) and small low-volume (1 to 5 l/min) PUFs available. PUF/XAD-2 "sandwiches" capture a wider range of semivolatiles.
Certifications: Compliant with EPA and ASTM methods—0.022 g/cm3 density.
Frequently Bought Together