Vacuum Bag Sampler, Restek
The model 1062 vacuum bag sampler provides fast sampling with zero cross-contamination.
- Fast bag sampling without sample passing through pump
- Bag capacity up to 10 L
- Observation window on case lid
- Sample inlet accepts 1/4" OD tubing
- Case designed for rugged outdoor use
A vacuum created in the box draws air into the sampling bag without drawing it through the vacuum pump first, as is the case with standard air sampling pumps, thereby preventing contamination of the sample. This bag sampler can fill a 10 L bag in two minutes with an automatic shut-off switch, which stops the sample bag from overfilling. The filling rate is adjusted with a vent rotometer valve. An external battery-recharging port enables continuous operation with battery charger. In addition, the quick exhaust valve allows for fast removal of the sampling bag.
Certifications: CE certified.
Ordering information: The sampler comes with a universal power adaptor/charger, battery, instruction manual, and 1-year limited warranty.
- Description:Sampling Pump, Vacuum Bag Sampler Model 1062, 1-10L gas sampling bag capacity
- Includes:Power adaptor, battery, manual