Order Entry
Canada
Orders LinkContactUsLinkComponent
Middlebrook OADC Supplement, Hardy Diagnostics

Middlebrook OADC Supplement, Hardy Diagnostics

Middlebrook OADC supplement, Bottle, 100 ml
Catalog # CA78658-128
New Product
Supplier #:  U98
Middlebrook OADC Supplement, Hardy Diagnostics
Catalog # CA78658-128
New Product
CAS Number:  
Middlebrook OADC Supplement, Hardy Diagnostics
Middlebrook OADC Supplement, Hardy Diagnostics

Middlebrook OADC supplement, Bottle, 100 ml

CAS Number:  
Price
$201.60
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

These supplements are ideal for isolation of Mycobacteria species.

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Middlebrook OADC supplement
  • Volume:
    100 ml
  • Cat. No.:
    CA78658-128
  • Storage temperature:
    2...8 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.