ContactUsLinkComponent
Seal Wash Plunger Seal for Waters HPLC Systems, Restek
Seal Wash Plunger Seal for Waters HPLC Systems, Restek
Catalog # 10853-586
Supplier:  Restek
undefined
Seal Wash Plunger Seal for Waters HPLC Systems, Restek
Catalog # 10853-586
Supplier:  Restek
Supplier Number:  25386
Specifications
  • Description:
    Seal Wash Plunger Seal for Waters Alliance
  • Cat. no.:
    10853-586
  • Model:
    2690, 2695, 2790, 2795
  • Supplier No.:
    25386