HPLC PEEK Performance Kit, Restek
Catalog # 10853-664
Supplier: Restek
Specifications
- Description:HPLC PEEK Performance Kit
- Cat. no.:10853-664
- Supplier No.:26394
Specifications
About this item
This HPLC PEEK Performance Kit is a useful complement to your HPLC system.
Ordering information: Includes Hub-Cap Adapter with Cap (1); Tubing Clips (3); Clean Cut Tubing Cutter (1); PEEK Finger Tight Fittings (5); PEEK Column Plugs (5); PEEK 90° Tubing Elbows (2); PEEK 180° Tubing Elbows (2); PEEK Column Connectors (2).