Outlet Cap and Gold Seal Assembly Tool for Agilent 1050, 1100 HPLC Systems, Restek
Catalog # 10853-792
Supplier: Restek
Specifications
- Description:Outlet Cap and Gold Seal Assembly Tool
- Cat. no.:10853-792
- Supplier No.:24989
Specifications
About this item
Easily install the gold seal into the outlet cap.
Ordering information: For use with Agilent 5001-3701 gold disk seal only. Not compatible with Restek cat.# 25889 and 25890 (Agilent 5067-4728).