Tubing Clip, Restek
Catalog #: 10059-412
Supplier: Restek
Specifications
- Description:Tubing Clip for 1/16" and 1/8" Tubing, 5-pk
- Cat. no.:10059-412
- Supplier No.:25310
Specifications
About this item
Securely holds 1/16" or 1/8" tubing in beaker, flask, or bottle up to 4 mm thick.
Customers Who Bought This Also Bought