Leukemia inhibitory factor (LIF) is a member of the interleukin 6 (IL-6) family that is made by a variety of adult and embryonic tissues. LIF signals through the glycoprotein 130 (gp130)/LIF receptor (LIFR) heterodimer to activate STAT3 and MAPK signaling. LIF functions during hematopoietic differentiation, neuronal cell differentiation, kidney development, and inflammatory processes. Mouse LIF promotes mouse embryonic stem (ES) cell self-renewal and pluripotency in long-term cell culture systems, similar to the functional activity of FGF-basic in human ES cell culture systems.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: IL-6 production by M1 cells, ED50 ≤1 ng/ml, Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P09056
- Reconstitution instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Leukocyte Inhibitory Factor
- Protein/peptide name:LIF
- Purity:≥95%
- Molecular weight:20 kDa
- Sequence:MSPLPITPVN ATCAIRHPCH GNLMNQIKNQ LAQLNGSANA LFISYYTAQG EPFPNNVEKL CAPNMTDFPS FHGNGTEKTK LVELYRMVAY LSASLTNITR DQKVLNPTAV SLQVKLNATI DVMRGLLSNV LCRLCNKYRV GHVDVPPVPD HSDKEAFQRK KLGCQLLGTY KQVISVVVQA F
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay