Human Recombinant PDGF-AB (Animal free) (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
Certificates
About this item
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells.
- High quality
- Low endotoxin
- Affordable
PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:−20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R-3T3 cell proliferation, ED50 ≤20 ng/ml, bioactivity ≥5.0×10⁴ units/mg
- Gene ID:P04085/P01127
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:ODGF|GDGF
- UniProtKB:P04085/P01127
- Protein/peptide name:PDGF-AB (AF)
- Purity:≥95%
- Molecular weight:Alpha: 14.4 kDa, beta: 12.4 kDa, total 26.8
- Sequence:Alpha chain: MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT Beta chain: MSLGSLTIAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRPTQVQLRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETVAAARPVT
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together