ContactUsLinkComponent
DIA-1000 Infecon Diagnostic Mailer, Com-pac
DIA-1000 Infecon Diagnostic Mailer, Com-pac
Catalog # 11217-084
Supplier:  Com-Pac
undefined
DIA-1000 Infecon Diagnostic Mailer, Com-pac
Catalog # 11217-084
Supplier:  Com-Pac
Supplier Number:  DIA-1000
Specifications
  • Description:
    DIA-1000 infecon diagnostic mailer
  • Cat. no.:
    11217-084
  • Supplier No.:
    DIA-1000
Specifications
About this item
This mailer consists of a 9" x 12" liquid-tight infecon bag with document pouch and biohazard symbol, one 6-tube bubble pouch, one 250 mL capacity absorbent sheet, and a sturdy shipping box.