Interleukin-19 (IL-19) is a member of the interleukin 10 (IL-10) cytokine family and is produced by B cells and monocytes. IL-19 binds the interleukin 20 receptor complex (IL-20R) to activate STAT3 signaling. IL-19 induces interleukin 6 (IL-6) and tumor necrosis factor alpha (TNF-a) expression in monocytes, and promotes T helper 2 (Th2) cell-mediated immune responses. IL-19 production is up-regulated in resting monocytes following granulocyte-macrophage colony-stimulating factor (GM-CSF) or lipopolysaccharide (LPS) stimulation.
- Animal-free
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q9UHD0
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Melanoma differentiation associated-like protein
- Protein/peptide name:IL-19
- Purity:≥95%
- Molecular weight:17.9 kDa
- Sequence:MLRRCLISTD MHHIEESFQE IKRAIQAKDT FPNVTILSTL ETLQIIKPLD VCCVTKNLLA FYVDRVFKDH QEPNPKILRK ISSIANSFLY MQKTLRQCQE QRQCHCRQEA TNATRVIHDN YDQLEVHAAA IKSLGELDVF LAWINKNHEV MFSA
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT