Order Entry
Canada
ContactUsLinkComponent
Human Transferrin, HRP (Horseradish Peroxidase)
Human Transferrin, HRP (Horseradish Peroxidase)
Catalog # CA009-0334
CAS Number:  
Human Transferrin, HRP (Horseradish Peroxidase)
Catalog # CA009-0334
Supplier Number:  009-0334
CAS Number:  

Specifications

  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Species:
    Human
  • Size:
    1 mg
  • Protein/peptide name:
    Transferrin
  • Cat. no.:
    CA009-0334
  • Supplier No.:
    009-0334

Specifications

About this item