Lab Burner
Catalog # 470019-572
Supplier: Wards
Specifications
- Description:Lab burner (supplied without gas cartridge)
- Environmentally Preferable:
- Cat. no.:470019-572
- Supplier No.:321201
Specifications
About this item
These laboratory burners are compact with a powerful and concentrated flame.
- Made of brass and stainless steel
Soot-free use and adjustable flame control. The burners screw into a cartridge to guarantee a leak free installation.
Burner has separate adjustments for regulating gas and air intake. Flame top: 1¼" diameter, burner unit: 2¼"×2⅔".