Order Entry
Canada
ContactUsLinkComponent
KIMBLE® ULTRA-WARE® 47 mm Microfiltration Assembly with GL 45 Style Bottle, DWK Life Sciences
KIMBLE® ULTRA-WARE® 47 mm Microfiltration Assembly with GL 45 Style Bottle, DWK Life Sciences
Catalog # KT953750-5347
undefined
KIMBLE® ULTRA-WARE® 47 mm Microfiltration Assembly with GL 45 Style Bottle, DWK Life Sciences
Catalog # KT953750-5347
Supplier Number:  953750-5347

Specifications

  • Filter Diameter:
    47 mm
  • Description:
    ULTRA-WARE® Microfiltration Assembly with GL 45 Style Bottle
  • Cat. no.:
    KT953750-5347
  • Supplier No.:
    953750-5347

Specifications

About this item

This filtration assembly is designed to collect filtrate directly into a media-style bottle.

  • All components are steam autoclavable
  • Sealed-in 40-60 micron porosity fritted glass disc
  • Vacuum and pressure rated from -1 to 1.5 bar
Certifications: Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Ordering information: This filtration assembly is supplied with funnel, clamp, fritted glass support base, tubing adapter, PBT cap, sealing ring and vacuum/pressure bottle.