ContactUsLinkComponent
Spec-Master® Series Stainless Steel Backsplash Work Table, 14-Gauge Type 300 Series, with Galvanized Steel Base and Undershelf, Eagle MHC™
Spec-Master® Series Stainless Steel Backsplash Work Table, 14-Gauge Type 300 Series, with Galvanized Steel Base and Undershelf, Eagle MHC™
Catalog # 58202-434
Supplier:  Eagle Group
undefined
Spec-Master® Series Stainless Steel Backsplash Work Table, 14-Gauge Type 300 Series, with Galvanized Steel Base and Undershelf, Eagle MHC™
Catalog # 58202-434
Supplier:  Eagle Group
Supplier Number:  T3072E-BS
Specifications
  • Description:
    E-BS Series Work Table
  • Cat. no.:
    58202-434
  • W×D:
    72×30"
  • Supplier No.:
    T3072E-BS
Specifications
About this item
14 Gauge, Type 300 series stainless steel top with 4½" backsplash at rear of table.
  • Rolled edge front with ends turned under 90 degrees
  • Adjustable working height (35⅛" to 36⅛")
1⅝" diameter legs and adjustable undershelf are heavy gauge galvanized steel.
Ordering information: Available accessories iinclude drawers and casters, sold separately.