Febrile Antigens and Antisera, Germaine® Laboratories
Supplier: Germaine Laboratories
Certificates
About this item
Febrile Antigens are intended for the detection of bacterial agglutinins in serum associated with a bacterial infection
The slide test is recommended as a screening procedure to establish the presence or absence of homologous antibodies. The tube test is intended to establish antibody titer. The Febrile Controls are intended for use as positive and negative controls for these tests.
Frequently Bought Together