Order Entry
Canada
ContactUsLinkComponent
Rhenium Standard for ICP, Certipur®, MilliporeSigma
Rhenium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70344.0100
Supplier:  MilliporeSigma
CAS Number:  
Rhenium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70344.0100
Supplier:  MilliporeSigma
Supplier Number:  1.70344.0100
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard class description:
    Corrosive Material
  • Cat. no.:
    CA1.70344.0100
  • pH:
    5 (H₂O, 20°C)
  • Element:
    Rhenium
  • Supplier No.:
    1.70344.0100

Specifications

About this item

NH₄ReO₄ in water

Traceable to SRM from NIST.

Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.