Order Entry
Canada
ContactUsLinkComponent
 
Praseodymium Standard for ICP, Certipur®, MilliporeSigma
  CA1.70343.0100
 :  
Praseodymium Standard for ICP, Certipur®, MilliporeSigma
  CA1.70343.0100
 :  1.70343.0100
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    2-3 % HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard class description:
    Corrosive Material
  • Cat. no.:
    CA1.70343.0100
  • pH:
    0.5 (H₂O, 20°C)
  • Element:
    Praseodymium
  • Supplier No.:
    1.70343.0100

 

 

Pr(NO₃)₃ in HNO₃

Traceable to SRM from NIST.

 : The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
 : Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.