Lithium Standard for ICP, Certipur®, MilliporeSigma
CA1.70329.0100
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Concentration:1000 µg/ml
- Matrix:2-3 % HNO₃
- Pack type:Plastic Bottle
- Size:100 ml
- Hazard class description:Corrosive Material
- Cat. no.:CA1.70329.0100
- pH:0.5 (H₂O, 20°C)
- Element:Lithium
- Supplier No.:1.70329.0100
Traceable to SRM from NIST.
: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.