Order Entry
Canada
ContactUsLinkComponent
 
Iridium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70325.0100
Supplier:  MilliporeSigma
CAS Number:  
Iridium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70325.0100
Supplier:  MilliporeSigma
Supplier Number:  1.70325.0100
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    7% HCl
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Hazard class description:
    Corrosive Material
  • Cat. no.:
    CA1.70325.0100
  • Element:
    Iridium
  • Supplier No.:
    1.70325.0100

Specifications

About this item

Traceable to SRM from NIST.

Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.