Order Entry
Canada
ContactUsLinkComponent
Cadmium Standard for ICP, Certipur®, MilliporeSigma
Cadmium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70309.0100
Supplier:  MilliporeSigma
CAS Number:  
Cadmium Standard for ICP, Certipur®, MilliporeSigma
Catalog # CA1.70309.0100
Supplier:  MilliporeSigma
Supplier Number:  1.70309.0100
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    2-3 % HNO₃
  • Pack type:
    Plastic Bottle
  • Size:
    100 ml
  • Cat. no.:
    CA1.70309.0100
  • Element:
    Cadmium
  • Supplier No.:
    1.70309.0100

Specifications

About this item

Traceable to SRM from NIST.

Certifications: The quality control of ICP single-element standards is carried out by the accredited calibration laboratory according to ISO 17025.
Ordering information: Includes a Certificate of Analysis, with exact data on content including uncertainty budget, trace element impurities, composition, traceability, date of release and minimum shelf life.