Order Entry
Canada
ContactUsLinkComponent
Sodium dodecyl sulfate (SDS), Supelco®
Sodium dodecyl sulfate (SDS), Supelco®
Catalog # CA1.18309.0025
Supplier:  MilliporeSigma
CAS Number:  
undefined
Sodium dodecyl sulfate (SDS), Supelco®
Catalog # CA1.18309.0025
Supplier:  MilliporeSigma
Supplier Number:  1.18309.0025
CAS Number:  

Specifications

  • Size:
    25 g
  • Environmentally Preferable:
  • Cat. no.:
    CA1.18309.0025
  • Supplier No.:
    1.18309.0025

Specifications

About this item

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
 :
 :288.38 g/mol
UN:1325
 :GHS02
 :GHS05
 :GHS07
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:>100 °C
ADR:4.1
Merck Index:14,08636