CTAB Extraction Solution, Teknova
About this item
CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.
The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20mM EDTA (pH 8,0) and 1.4M Sodium Chloride.
1 Options Available Below
Product Details & Documents
CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.
The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20mM EDTA (pH 8,0) and 1.4M Sodium Chloride.
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- SizeAvailabilityPrice
- CA77443-472CTAB extraction solution, sterileSupplier Number: C2190Specifications:New ProductNew Product
- CA100219-610CTAB extraction solution, sterileCTAB extraction solution, sterile250 mlDiscontinued-Supplier Number: -