To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:LabSafe™ Nucleic Acid Stain (10,000X)
- Size:1 ml
- Cat. no.:CA89233-908
- Supplier No.:786-409
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels.
- Non-carcinogenic by the Ames-test
- Used commonly in gel electrophoresis for visualization of nucleic acids
- Replace toxic Ethidium Bromide (a potent mutagen)
LabSafe™ Nucleic Acid Stain is a safe nucleic acid stain for the visualization of double-stranded DNA, single-stranded DNA, and RNA in agarose gels. The results are negative in both the mouse marrow chromophilous erythrocyte micronucleus and mouse primary spermatocyte chromosomal aberration tests.