You searched for: Serological Pipettes
Reusable glass or disposable polystyrene serological pipettes aspirate and release milliliter liquid amounts using the blow-out method. The narrow tube construction is transparent and may come with or without calibrated graduations in various scales for accurate meniscus measurements. These volumetric transfer instruments come sterile for aseptic cell culturing procedures. Choose between non-plugged or cotton plug options to prevent overfilling. Color-coded for correct size identification, serological pipettes are available in different lengths and capacities.
VWR® Serological Pipette
Supplier: VWR International
These serological plastic pipettes are designed to use with tissue culture work.
Expand 1 Items
VWR® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged
Supplier: VWR International
VWR® Disposable Serological Pipettes are manufactured from polystyrene and are supplied sterile and plugged.
Expand 1 Items
VWR® Open End Serological Pipettes
Supplier: VWR International
Wide, open end design is ideal for viscous and high particulate solutions, as well as slurries such as Sephadex.
Expand 8 Items
Wobble-not: Serological Pipets Reimagined
Patented innovative 2-tier step design at the plug. Maximum stability for less dripping, more accuracy, and less risk of contamination.
VWR® Disposable Serological Pipettes
Supplier: VWR International
Polystyrene, transparent and graduated.
Expand 43 Items
VWR® Disposable Aspirating Pipettes, Polystyrene, Sterile
Supplier: VWR International
VWR® Disposable Aspirating Pipettes are manufactured from polystyrene and supplied sterile.
Expand 5 Items
VWR® Volumetric Pipettes, Reusable, Color Coded, Class A
Supplier: VWR International
Made from ASTM E438 Type 1 Class A borosilicate 33 expansion glass and as per ASTM specification E969.
Expand 18 Items
VWR® Disposable Serological Pipettes, Glass
Supplier: VWR International
VWR® Disposable Serological Pipettes are available in sterile or nonsterile formats, with a variety of packaging options to choose from.
Expand 19 Items
Corning® Stripette® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
PS, transparent, graduated, sterile.
Expand 29 Items
PYREX® Serological Pipettes, Plugged
Supplier: Corning
These sterile, glass pipettes, calibrated to deliver with blow-out; offer long slender tapered tips to make pipetting faster and easier.
Expand 5 Items
PYREX® Serological Pipettes, Corning
Supplier: Corning
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
Expand 26 Items
PYREX® Reusable Serological Pipettes, Glass, Corning
Supplier: Corning
Reusable serological pipettes made of PYREX® glass.
Expand 8 Items
Shorty Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
Disposable polystyrene shorty pipettes are 11.4 cm (4 ¹/₂") shorter than the standard length serological pipettes, making them ideal for working under benchtop hoods while significantly reducing hand/wrist fatigue.
Expand 2 Items
Nunc™ Serological Pipettes, Thermo Scientific
Supplier: Thermo Fisher Scientific
Select size and packaging options to advance your research with complete confidence when using Nunc™ Serological Pipettes
Expand 18 Items
Disposable Serological Pipettes, Polystyrene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
Pipettes are manufactured from optical-grade polystyrene and feature a high internal and external surface polish, which reduces meniscus formation and helps ensure complete liquid expulsion.
Expand 24 Items
Disposable Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
These accurate, plastic pipettes are sterile, plugged, and ready for use.
Expand 8 Items
Disposable Standard-Tip Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Standard-Tip Serological Pipettes are irradiation sterilized and suitable for tissue culture work.
Expand 10 Items
Disposable Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
Argos brand standard serological pipettes are designed for accurate and rapid fluid dispensing
Expand 29 Items
Disposable Mini Plastic Serological Pipettes, Argos Technologies
Supplier: Argos Technologies
Argos brand mini pipettes were developed for use in confined spaces such as laminar hoods
Expand 3 Items
Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
These pipettes feature a uniform pulled tip and are clean, plugged, and ready for use.
Expand 10 Items
Disposable Serological Pipettes, Glass, Sterile, TD Color-Coded, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements.
Expand 4 Items
Dye Pipettes, Glass, Color-Coded
Supplier: DWK Life Sciences (KIMBLE)
Calibrated to deliver with blowout.
Expand 4 Items
Shorty Disposable Serological Pipettes, Glass, Sterile, Plugged, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Calibrated to deliver with blowout after free flow of liquid has ended
Expand 10 Items
KIMBLE® Sterile Disposable Glass, Wide Tip Serological Pipette, Plugged
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Wide tips are excellent for transferring viscous or suspended liquids.
Expand 2 Items
Disposable Serological Pipettes, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Pipettes are non-plugged and nonsterile, with a uniform pulled tip.
Expand 5 Items
Thermo Scientific™ Serological Pipettes
Supplier: Thermo Fisher Scientific
Bold, clear bi-directional graduations make it easy-to-read fluid volumes and dispensing amounts. Choose from a broad selection of sizes and packaging options.
Expand 18 Items
Serological Pipet, Shortie, Thermo Scientific
Supplier: Thermo Fisher Scientific
Plugged and individually wrapped in paper peel packaging to maintain sterility.
Expand 2 Items
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Wide-tip pipettes with large tip opening provide faster delivery and are ideal for use with viscous liquids, slurries, and suspensions
Expand 4 Items
SP Wilmad-LabGlass Serological Color Coded Class B Pipettes, SP Industries
Supplier: SP Industries
Calibrated 'to deliver' (TD) at 20°C, these borosilicate glass pipettes have a permanently colored amber scale with the zero line at the top
Expand 2 Items
KIMAX® Reusable Serological Pipettes, Glass, Color-Coded, Kimble Chase
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
These KIMAX-51® borosilicate glass pipettes are intended for bacteriological, microbiological, or serological work. Pipettes must be blown out after free delivery to obtain the total rated capacity. Scale is permanent brown stain fused into the surface of uniform bore tubing without etching. Plain, fire-polished top end has been tooled to accept cotton plugging. Color-coding facilitates sorting and selection. Designed from ASTM Specification E1044, Style II requirements.
Expand 9 Items
Falcon® Disposable Serological Pipettes, Polystyrene, Sterile, Plugged, Corning
Supplier: Corning
Color-coded pipettes feature both negative and reverse graduations (except on 1 ml pipet).