Protexis™ PI Micro Sterile Polyisoprene Powder-Free Surgical Gloves, Cardinal Health
Supplier: Cardinal Health
Certificates
About this item
Surgical, examination gloves with a unique synthetic polyisoprene formulation, identical to natural rubber latex, without the proteins.
- Thinner for more tactile sensitivity
- Proprietary glove mold
- Interlocking cuff style
These gloves offer healthcare works an alternative solution that performs like latex in terms of comfort, fit and feel.
Exclusive glove mold with independent thumb design that allows for an anatomical fit and natural movement in the fingers, thumb and palm. Proprietary interlocking, beaded cuff design reduces roll-down.
Helps to reduce powder-related skin irritation and post-operative complications associate with granulomas. 20% thinner than Protexis™ PI Surgical Gloves for an enhanced tactile response. Excellent choice as an outer glove when double-gloving. Produced on Cardinal Health™ Surgical Glove Molds.
Certifications: Tested in accordance with ASTM D 6124.
Packaging: 50 pairs/box, 4 boxes/case.
Specifications
- Catalog No:
- 89411-644
- 89411-642
- 89411-640
- 89411-650
- 89411-648
- 89411-646
- 89411-654
- 89411-652
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded