Platinum Dishes, Heraeus
Supplier: HERAEUS MATERIALS TECHNOLOGY NA
Certificates
About this item
These Dishes are made with 99.95% Pure Platinum.
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Dishes have become essential for the preparation of samples in analytical laboratories.
Ordering information: Pourling Lip available upon request. Lids sold separately. For additional lid details, see Lids for Platinum and Platinum 95%/Gold 5% Dishes, Heraeus.
Customers Who Bought This Also Bought