Platinum Crucibles, Heraeus
Supplier: HERAEUS MATERIALS TECHNOLOGY NA
Certificates
About this item
These Crucibles are made with 99.95% Pure Platinum
Due to their resistance to oxidation and corrosion, as well as mechanical stability at high temperatures, Platinum Crucibles have become essential for the preparation of samples in analytical laboratories.
They are available in Standard Form or Wide Form.
Ordering information: Lids sold separately. For additional lid details, see Lids for Platinum and Platinum 95%/Gold 5% Crucibles, Heraeus.
Customers Who Bought This Also Bought